Merck
  • Critical involvement of pneumolysin in production of interleukin-1alpha and caspase-1-dependent cytokines in infection with Streptococcus pneumoniae in vitro: a novel function of pneumolysin in caspase-1 activation.
Critical involvement of pneumolysin in production of interleukin-1alpha and caspase-1-dependent cytokines in infection with Streptococcus pneumoniae in vitro: a novel function of pneumolysin in caspase-1 activation.
Infection and immunity (2008-01-16)
Shereen Shoma, Kohsuke Tsuchiya, Ikuo Kawamura, Takamasa Nomura, Hideki Hara, Ryosuke Uchiyama, Sylvia Daim, Masao Mitsuyama
ABSTRACT
Pneumolysin is a pore-forming cytolysin known as a major virulence determinant of Streptococcus pneumoniae. This protein toxin has also been shown to activate the Toll-like receptor 4 (TLR4) signaling pathway. In this study, a mutant S. pneumoniae strain deficient in pneumolysin (Deltaply) and a recombinant pneumolysin protein (rPLY) were constructed. Upon infection of macrophages in vitro, the ability to induce the production of interleukin-1alpha (IL-1alpha), IL-1beta, and IL-18 was severely impaired in the Deltaply mutant, whereas there was no marked difference in the induction of tumor necrosis factor alpha (TNF-alpha) and IL-12p40 between the wild type and the Deltaply mutant of S. pneumoniae. When macrophages were stimulated with rPLY, the production of IL-1alpha, IL-1beta, and IL-18 was strongly induced in a TLR4-dependent manner, whereas lipopolysaccharide, a canonical TLR4 agonist, hardly induced these cytokines. In contrast, lipopolysaccharide was more potent than rPLY in inducing the production of TNF-alpha, IL-6, and IL-12p40, the cytokines requiring no caspase activation. Activation of caspase-1 was observed in macrophages stimulated with rPLY but not in those stimulated with lipopolysaccharide, and the level of activation was higher in macrophages infected with wild-type S. pneumoniae than in those infected with the Deltaply mutant. These results clearly indicate that pneumolysin plays a key role in the host response to S. pneumoniae, particularly in the induction of caspase-1-dependent cytokines.
Sigma-Aldrich
Lipopolysaccharides from Escherichia coli O55:B5, Ready Made solution, 1 mg/mL
Sigma-Aldrich
Lipopolysaccharides from Escherichia coli O55:B5, FITC conjugate
Sigma-Aldrich
Lipopolysaccharides from Escherichia coli O55:B5, purified by gel-filtration chromatography
Sigma-Aldrich
Lipopolysaccharides from Escherichia coli O55:B5, purified by ion-exchange chromatography, TLR ligand tested
Sigma-Aldrich
Lipopolysaccharides from Escherichia coli O55:B5, purified by phenol extraction
Sigma-Aldrich
Lipopolysaccharides from Escherichia coli O55:B5, purified by trichloroacetic acid extraction
Sigma-Aldrich
Lipopolysaccharides from Escherichia coli O55:B5, γ-irradiated, BioXtra, suitable for cell culture