Merck
  • The malaria vector mosquito Anopheles gambiae expresses a suite of larval-specific defensin genes.
The malaria vector mosquito Anopheles gambiae expresses a suite of larval-specific defensin genes.
Insect molecular biology (2008-03-21)
J M Meredith, H Hurd, M J Lehane, P Eggleston
ABSTRACT
cDNAs of Anopheles gambiae Defensin 2 (AgDef2), Defensin 3 (AgDef3) and Defensin 4 (AgDef4), identified in the genome sequence, have been characterized and their expression profiles investigated. In contrast to both typical defensins and insect antimicrobial peptides generally, the newly identified defensins were not upregulated with acute-phase kinetics following immune challenge in insects or cell culture. However, mRNA abundance of AgDef2, AgDef3 and AgDef4 increased significantly during the larval stages. Promoter analysis of all three genes failed to identify putative immune response elements previously identified in other mosquito defensin genes. As previous studies failed to identify these larval-specific defensins, it seems likely that further antimicrobial peptide genes with nontypical expression profiles will be identified as more genome sequences become available.
Sigma-Aldrich
GenElute Mammalian Genomic DNA Miniprep Kits, sufficient for 350 purifications
Sigma-Aldrich
GenElute Mammalian Genomic DNA Miniprep Kits, sufficient for 70 purifications
Sigma-Aldrich
GenElute Mammalian Genomic DNA Miniprep Kits, sufficient for 10 purifications
Sigma-Aldrich
20-Hydroxyecdysone, ≥93% (HPLC), powder
Sigma-Aldrich
Laminarin from Laminaria digitata, polysaccharide substrate for laminarinase