Merck
  • Transcriptional and epigenetic dysregulation impairs generation of proliferative neural stem and progenitor cells during brain aging.
Transcriptional and epigenetic dysregulation impairs generation of proliferative neural stem and progenitor cells during brain aging.
Nature aging (2024-01-05)
Meiyang Li, Hongzhi Guo, Michael Carey, Chengyang Huang
ABSTRACT
The decline in stem cell function during aging may affect the regenerative capacity of mammalian organisms; however, the gene regulatory mechanism underlying this decline remains unclear. Here we show that the aging of neural stem and progenitor cells (NSPCs) in the male mouse brain is characterized by a decrease in the generation efficacy of proliferative NSPCs rather than the changes in lineage specificity of NSPCs. We reveal that the downregulation of age-dependent genes in NSPCs drives cell aging by decreasing the population of actively proliferating NSPCs while increasing the expression of quiescence markers. We found that epigenetic deregulation of the MLL complex at promoters leads to transcriptional inactivation of age-dependent genes, highlighting the importance of the dynamic interaction between histone modifiers and gene regulatory elements in regulating transcriptional program of aging cells. Our study sheds light on the key intrinsic mechanisms driving stem cell aging through epigenetic regulators and identifies potential rejuvenation targets that could restore the function of aging stem cells.
Sigma-Aldrich
Anti-MLL4 Antibody, from rabbit, purified by affinity chromatography
Sigma-Aldrich
Anti-HP1γ Antibody, clone 42s2, clone 42s2, Upstate®, from mouse