Merck
  • Characterization of the sulfhydryl-sensitive site in the enzyme responsible for hydrolysis of 2-arachidonoyl-glycerol in rat cerebellar membranes.
Characterization of the sulfhydryl-sensitive site in the enzyme responsible for hydrolysis of 2-arachidonoyl-glycerol in rat cerebellar membranes.
Chemistry & biology (2005-06-25)
Susanna M Saario, Outi M H Salo, Tapio Nevalainen, Antti Poso, Jarmo T Laitinen, Tomi Järvinen, Riku Niemi
ABSTRACT
We have previously reported that the endocannabinoid, 2-arachidonoyl-glycerol (2-AG), is hydrolyzed in rat cerebellar membranes by monoglyceride lipase (MGL)-like enzymatic activity. The present study shows that, like MGL, 2-AG-degrading enzymatic activity is sensitive to inhibition by sulfhydryl-specific reagents. Inhibition studies of this enzymatic activity by N-ethylmaleimide analogs revealed that analogs with bulky hydrophobic N-substitution were more potent inhibitors than hydrophilic or less bulky agents. Interestingly, the substrate analog N-arachidonylmaleimide was found to be the most potent inhibitor. A comparison model of MGL was constructed to get a view on the cysteine residues located near the binding site. These findings support our previous conclusion that the 2-AG-degrading enzymatic activity in rat cerebellar membranes corresponds to MGL or MGL-like enzyme and should facilitate further efforts to develop potent and more selective MGL inhibitors.