Merck
  • Identification of a region of beta-amyloid precursor protein essential for its gelatinase A inhibitory activity.
Identification of a region of beta-amyloid precursor protein essential for its gelatinase A inhibitory activity.
The Journal of biological chemistry (2003-02-15)
Shouichi Higashi, Kaoru Miyazaki
ABSTRACT
Because beta-amyloid precursor protein (APP) has the abilities both to interact with extracellular matrix and to inhibit gelatinase A activity, this molecule is assumed to play a regulatory role in the gelatinase A-catalyzed degradation of extracellular matrix. To determine a region of APP essential for the inhibitory activity, we prepared various derivatives of APP. Functional analyses of proteolytic fragments of soluble APP (sAPP) and glutathione S-transferase fusion proteins, which contain various COOH-terminal parts of sAPP, showed that a site containing residues 579-601 of APP(770) is essential for the inhibitory activity. Moreover, a synthetic decapeptide containing the ISYGNDALMP sequence corresponding to residues 586-595 of APP(770) had a gelatinase A inhibitory activity slightly higher than that of sAPP. Studies of deletion of the NH(2)- and COOH-terminal residues and alanine replacement of internal residues of the decapeptide further revealed that Tyr(588), Asp(591), and Leu(593) of APP mainly stabilize the interaction between gelatinase A and the inhibitor. We also found that the residues of Ile(586), Met(594), and Pro(595) modestly contribute to the inhibitory activity. The APP-derived decapeptide efficiently inhibited the activity of gelatinase A (IC(50) = 30 nm), whereas its inhibitory activity toward membrane type 1 matrix metalloproteinase was much weaker (IC(50) = 2 microm). The decapeptide had poor inhibitory activity toward gelatinase B, matrilysin, and stromelysin (IC(50) > 10 microm). The APP-derived inhibitor formed a complex with active gelatinase A but not with progelatinase A, and the complex formation was prevented completely by a hydroxamate-based synthetic inhibitor. Therefore, the decapeptide region of APP is likely an active site-directed inhibitor that has high selectivity toward gelatinase A.
Sigma-Aldrich
Achromopeptidase from bacteria, partially purified powder, ≥20,000 units/mg solid