Merck
  • Respiratory and TCA cycle activities affect S. cerevisiae lifespan, response to caloric restriction and mtDNA stability.
Respiratory and TCA cycle activities affect S. cerevisiae lifespan, response to caloric restriction and mtDNA stability.
Journal of bioenergetics and biomembranes (2011-08-13)
Erich B Tahara, Kizzy Cezário, Nadja C Souza-Pinto, Mario H Barros, Alicia J Kowaltowski
ABSTRACT
We studied the importance of respiratory fitness in S. cerevisiae lifespan, response to caloric restriction (CR) and mtDNA stability. Mutants harboring mtDNA instability and electron transport defects do not respond to CR, while tricarboxylic acid cycle mutants presented extended lifespans due to CR. Interestingly, mtDNA is unstable in cells lacking dihydrolipoyl dehydrogenase under CR conditions, and cells lacking aconitase under standard conditions (both enzymes are components of the TCA and mitochondrial nucleoid). Altogether, our data indicate that respiratory integrity is required for lifespan extension by CR and that mtDNA stability is regulated by nucleoid proteins in a glucose-sensitive manner.
Sigma-Aldrich
Diaphorase from Clostridium kluyveri, recombinant, expressed in E. coli, Animal-component free
Sigma-Aldrich
Diaphorase from Clostridium kluyveri, lyophilized powder, 3.0-20.0 units/mg protein (biuret)