Merck
  • Lipid-bound apolipoproteins in tyrosyl radical-oxidized HDL stabilize ABCA1 like lipid-free apolipoprotein A-I.
Lipid-bound apolipoproteins in tyrosyl radical-oxidized HDL stabilize ABCA1 like lipid-free apolipoprotein A-I.
BMC biochemistry (2012-01-18)
Mohammad A Hossain, Sereyrath Ngeth, Teddy Chan, Michael N Oda, Gordon A Francis
ABSTRACT
ATP-binding cassette transporter A1 (ABCA1) mediates the lipidation of exchangeable apolipoproteins, the rate-limiting step in the formation of high density lipoproteins (HDL). We previously demonstrated that HDL oxidized ex vivo by peroxidase-generated tyrosyl radical (tyrosylated HDL, tyrHDL) increases the availability of cellular cholesterol for efflux and reduces the development of atherosclerosis when administered to apolipoprotein E-deficient mice as compared to treatment with control HDL. In the current study we determined that tyrHDL requires functional ABCA1 for this enhanced activity. Like lipid-free apolipoprotein A-I (apoA-I), tyrHDL increases total and cell surface ABCA1, inhibits calpain-dependent and -independent proteolysis of ABCA1, and can be bound by cell surface ABCA1 in human skin fibroblasts. Additionally, tyrHDL apoproteins are susceptible to digestion by enteropeptidase like lipid-free apoA-I, but unlike lipid-bound apoA-I on HDL, which is resistant to proteolysis. These results provide the first evidence that lipid-bound apolipoproteins on the surface of spherical HDL particles can behave like lipid-free apoA-I to increase ABCA1 protein levels and activity.
Sigma-Aldrich
Enterokinase from porcine intestine, ≥0.5 units/mg solid
Sigma-Aldrich
Enterokinase from porcine intestine, lyophilized powder, ≥100 units/mg protein
Sigma-Aldrich
Anti-Apolipoprotein A-I Rabbit pAb, liquid, Calbiochem®
Sigma-Aldrich
Enterokinase from bovine intestine, powder