Merck
  • Biological evaluation of RBx-0128, a potent and selective dipeptidyl peptidase-IV inhibitor in type 2 diabetes genetic model.
Biological evaluation of RBx-0128, a potent and selective dipeptidyl peptidase-IV inhibitor in type 2 diabetes genetic model.
Indian journal of pharmacology (2012-12-19)
Joseph A Davis, Pucha S Kumar, Shuchita Singh, A Surender, Subhasis Roy, Vivek Khanna, Sachin Sethi, Chanchan Pal, Lalima Sharma, Biju Benjamin, Shivani Mittra, Jitendra Sattigeri, Vinay S Bansal
ABSTRACT
Dipeptidyl peptidase IV (DPP-IV) inhibition to modulate the incretin effect is a proven strategy to treat type 2 diabetes mellitus. The present study describes the pharmacological profile of a novel DPP-IV inhibitor RBx-0128, as an antidiabetic agent. DPP-IV assay was carried out to evaluate in vitro potency of RBx-0128 using human, mouse, and rat plasma as an enzyme source. Selectivity was assessed with various serine proteases. In vivo efficacy was assessed in ob/ob mice. The pharmacokinetic (PK) profile was performed in Wistar rats. RBx-0128 inhibited human, mouse, and rat plasma DPP-IV activity with IC50 values of 10.6, 18.1, and 56.0 nM respectively, selective over various serine proteases (900-9000-fold). The inhibition was reversible and competitive in nature. In ob/ob mice, RBx-0128 significantly (P<0.05) inhibited plasma DPP-IV and stimulated GLP-1 and insulin at 10 mg/kg. In the oral glucose tolerance test (OGTT), glucose lowering effect was better than sitagliptin (23 vs. 17%) at 10 mg/kg. The effect was sustained till 8 hours (30-35%) at 10 mg/kg with favorable PK profile (plasma clearance: 39.3 ml/min/kg; Cmax 790 ng/ml; t1/2 1.6 hours; tmax 4.8 hours, Vss 3.24 l/kg and Foral 55%) in Wistar rats. The present study showed that RBx-0128 is a novel, DPP-IV inhibitor with an antihyperglycemic effect. It can be a promising candidate for the treatment of type 2 diabetes mellitus.
Sigma-Aldrich
1-Ethyl-2-pyrrolidone, 98%
Sigma-Aldrich
Dipeptidyl Peptidase IV human, recombinant, expressed in Sf9 cells
Sigma-Aldrich
Dipeptidyl Peptidase IV human, recombinant, expressed in baculovirus infected Sf9 cells, pkg of ≥1.0 units/vial, ≥10 units/mg protein
Sigma-Aldrich
Phe-Pro
Sigma-Aldrich
N-Succinyl-Ala-Ala-Ala-p-nitroanilide, elastase substrate