Merck
  • Discovery, biological evaluation, and structure-activity and -selectivity relationships of 6'-substituted (E)-2-(benzofuran-3(2H)-ylidene)-N-methylacetamides, a novel class of potent and selective monoamine oxidase inhibitors.
Discovery, biological evaluation, and structure-activity and -selectivity relationships of 6'-substituted (E)-2-(benzofuran-3(2H)-ylidene)-N-methylacetamides, a novel class of potent and selective monoamine oxidase inhibitors.
Journal of medicinal chemistry (2013-02-27)
Leonardo Pisani, Maria Barletta, Ramon Soto-Otero, Orazio Nicolotti, Estefania Mendez-Alvarez, Marco Catto, Antonellina Introcaso, Angela Stefanachi, Saverio Cellamare, Cosimo Altomare, Angelo Carotti
ABSTRACT
The use of selective inhibitors of monoamine oxidase A (MAO-A) and B (MAO-B) holds a therapeutic relevance in the treatment of depressive disorders and Parkinson's disease (PD), respectively. Here, the discovery of a new class of compounds acting as monoamine oxidase inhibitors (MAO-Is) and bearing a 6'-substituted (E)-2-(benzofuran-3(2H)-ylidene)-N-alkylacetamide skeleton is reported. 6'-Sulfonyloxy derivatives exhibited outstanding affinities to MAO-A (7.0 nM < IC50 < 49 nM, much higher than moclobemide) and a pronounced MAO-A/B selectivity. The corresponding 6'-benzyloxy derivatives showed potent MAO-B inhibition and inverted selectivity profile. The rigid E-geometry of the exocyclic double bond allowed a more efficient binding conformation compared to more flexible and less active 2-(1-benzofuran-3-yl)-N-methylacetamide isomers and 4-N-methylcarboxamidomethylcoumarin analogues. Focused structural modifications and docking simulations enabled the identification of key molecular determinants for high affinity toward both MAO isoforms. These novel MAO-Is may represent promising hits for the development of safer therapeutic agents with a potential against depression, PD, and other age-related neurodegenerative pathologies.
Sigma-Aldrich
Monoamine Oxidase A human, recombinant, expressed in baculovirus infected BTI insect cells
Sigma-Aldrich
2,3-Benzofuran, 99%
Sigma-Aldrich
N-(Diphenylmethyl)methylamine, ≥98.0% (GC)
Sigma-Aldrich
Monoamine Oxidase B human, recombinant, expressed in baculovirus infected BTI insect cells