Merck
  • Vorapaxar in patients with peripheral artery disease: results from TRA2{degrees}P-TIMI 50.
Vorapaxar in patients with peripheral artery disease: results from TRA2{degrees}P-TIMI 50.
Circulation (2013-03-19)
Marc P Bonaca, Benjamin M Scirica, Mark A Creager, Jeffrey Olin, Henri Bounameaux, Mikael Dellborg, Jessica M Lamp, Sabina A Murphy, Eugene Braunwald, David A Morrow
ABSTRACT
Vorapaxar is a novel antagonist of protease-activated receptor-1, the primary receptor for thrombin on human platelets that is also present on vascular endothelium and smooth muscle. Patients with peripheral artery disease are at risk of systemic atherothrombotic events, as well as acute and chronic limb ischemia and the need for peripheral revascularization. The Trial to Assess the Effects of SCH 530348 in Preventing Heart Attack and Stroke in Patients With Atherosclerosis (TRA2°P-TIMI 50) was a randomized, double-blind, placebo-controlled trial of vorapaxar in 26 449 patients with stable atherosclerotic vascular disease (myocardial infarction, stroke, or peripheral artery disease). Patients with qualifying peripheral artery disease (n=3787) had a history of claudication and an ankle-brachial index of <0.85 or prior revascularization for limb ischemia. The primary efficacy end point was cardiovascular death, myocardial infarction, or stroke, and the principal safety end point was Global Utilization of Streptokinase and t-PA for Occluded Coronary Arteries (GUSTO) bleeding. In the peripheral artery disease cohort, the primary end point did not differ significantly with vorapaxar (11.3% versus 11.9%; hazard ratio, 0.94; 95% confidence interval, 0.78-1.14; P=0.53). However, rates of hospitalization for acute limb ischemia (2.3% versus 3.9%; hazard ratio, 0.58; 95% confidence interval, 0.39-0.86; P=0.006) and peripheral artery revascularization (18.4% versus 22.2%; hazard ratio, 0.84; 95% confidence interval, 0.73-0.97; P=0.017) were significantly lower in patients randomized to vorapaxar. Bleeding occurred more frequently with vorapaxar compared with placebo (7.4% versus 4.5%; hazard ratio, 1.62; 95% confidence interval, 1.21-2.18; P=0.001). Vorapaxar did not reduce the risk of cardiovascular death, myocardial infarction, or stroke in patients with peripheral artery disease; however, vorapaxar significantly reduced acute limb ischemia and peripheral revascularization. The beneficial effects of protease-activated receptor-1 antagonism on limb vascular events were accompanied by an increased risk of bleeding.
Sigma-Aldrich
Thrombin human, BioUltra, recombinant, expressed in HEK 293 cells, aqueous solution, ≥95% (SDS-PAGE)
Sigma-Aldrich
Thrombin from human plasma, 400-1000 NIH units/mg protein
Sigma-Aldrich
Thrombin from human plasma, lyophilized powder, 1500-3500 NIH units/mg protein (E1%/280, 18.3), suitable for cell culture
Sigma-Aldrich
Thrombin from human plasma, lyophilized powder, ≥2,000 NIH units/mg protein (E1%/280, 18.3)
Sigma-Aldrich
Thrombin from human plasma, lyophilized powder, ≥1,000 NIH units/mg protein (E1%/280, 18.3)
Sigma-Aldrich
Thrombin from human plasma, lyophilized powder, Suitable for routine use in the thrombin time test
Sigma-Aldrich
Thrombin from human plasma, lyophilized powder, ≥2800 NIH units/mg protein (E1%/280, 18.3)
Sigma-Aldrich
Thrombin from bovine plasma, ≥60 NIH units/mg protein (biuret)
Sigma-Aldrich
Thrombin from bovine plasma, lyophilized powder, 40-500 NIH units/mg protein (biuret)
Sigma-Aldrich
Thrombin from bovine plasma, lyophilized powder, ≥2,000 NIH units/mg protein (E1%/280 = 19.5)
Sigma-Aldrich
Thrombin from bovine plasma, lyophilized powder, 600-2,000 NIH units/mg protein (biuret)