Merck
  • Nontypeable Haemophilus influenzae carbonic anhydrase is important for environmental and intracellular survival.
Nontypeable Haemophilus influenzae carbonic anhydrase is important for environmental and intracellular survival.
Journal of bacteriology (2013-04-09)
Jeroen D Langereis, Aldert Zomer, Hendrik G Stunnenberg, Peter Burghout, Peter W M Hermans
ABSTRACT
Nontypeable Haemophilus influenzae (NTHi) is one of the leading causes of noninvasive mucosal infections, such as otitis media, sinusitis, and conjunctivitis. During its life cycle, NTHi is exposed to different CO2 levels, which vary from ∼0.04% in ambient air during transmission to a new host to over 5% in the respiratory tract and tissues of the human host during colonization and disease. We used the next-generation sequencing Tn-seq technology to identify genes essential for NTHi adaptation to changes in environmental CO2 levels. It appeared that H. influenzae carbonic anhydrase (HICA), which catalyzes the reversible hydration of CO2 to bicarbonate, is a molecular factor that is conditionally essential for NTHi survival in ambient air. Growth of NTHi Δcan strains was restored under 5% CO2-enriched conditions, by supplementation of the growth medium with sodium bicarbonate, or by genetic complementation with the can gene. Finally, we showed that HICA not only is essential for environmental survival but also appeared to be important for intracellular survival in host cells. Hence, HICA is important for NTHi niche adaptation.
Sigma-Aldrich
Carbonic Anhydrase from bovine erythrocytes, ≥95% (SDS-PAGE), specific activity ≥3,500 W-A units/mg protein, lyophilized powder
Sigma-Aldrich
Carbonic Anhydrase from bovine erythrocytes, lyophilized powder, ≥2,000 W-A units/mg protein
Sigma-Aldrich
Carbonic Anhydrase I from human erythrocytes
Sigma-Aldrich
Carbonic Anhydrase from bovine erythrocytes, BioReagent, suitable for GFC marker
Sigma-Aldrich
Carbonic Anhydrase from bovine erythrocytes, For use as a marker in SDS-PAGE
Sigma-Aldrich
Carbonic Anhydrase from bovine erythrocytes, non-denaturing PAGE marker
Supelco
Carbonic Anhydrase I from human erythrocytes, Isoelectric focusing marker, pI 6.6
Sigma-Aldrich
Carbonic Anhydrase Isozyme II from bovine erythrocytes, lyophilized powder, ≥3,000 W-A units/mg protein
Sigma-Aldrich
Carbonic Anhydrase Isozyme II from bovine erythrocytes, lyophilized powder, ≥2,000 W-A units/mg protein
Sigma-Aldrich
Carbonic Anhydrase Isozyme II human, ≥80%, powder, ≥3,000 W-A units/mg protein
Sigma-Aldrich
Carbonic Anhydrase Isozyme II from bovine erythrocytes, Isoelectric focusing marker, pI 5.4
Sigma-Aldrich
Carbonic Anhydrase II bovine, ≥90% (SDS-PAGE), recombinant, expressed in E. coli, ≥5,000 units/mg protein, buffered aqueous solution
Sigma-Aldrich
Carbonic Anhydrase II human, recombinant, expressed in E. coli, buffered aqueous solution