Merck
  • Blood lipid peroxidation, antioxidant enzyme activities and hemorheological changes in autistic children.
Blood lipid peroxidation, antioxidant enzyme activities and hemorheological changes in autistic children.
Ideggyogyaszati szemle (2013-04-24)
Aranka László, Zoltán Novák, Ilona Szőllősi-Varga, Du Quai Hai, Ágnes Vetró, Attila Kovács
ABSTRACT
Early infantile autism is a severe form of childhood psychiatric disease with characteristic symptoms. Hyperserotoninaemia in 43.5%, lactic acidosis 43% and hyperpyruvataemia in 30% were biochemically demonstrated in autistic children. Our earlier results led to the postulation that a dissequilibrium in the blood redox is involved in infantile autism; the oxidative loading and the antioxidant defending enzyme system were investigated together with the hemorheological parameters in infantile autism. Malonyl-dialdehyde (MDA) endproduct of lipid peroxidation and activities of the antioxidant enzymes: superoxide dismutase (SOD), catalase (C-ase), glutathione peroxidase (GP-ase) and reduced glutathione (GSH) were biochemically determined from plasma and red blood cells. The antioxidant specificities were investigated in plasma and red blood cell haemolysate from 25 infantile autistic children. Significantly increased superoxide dismutase (SOD) (2.89 vs. 1.32 U/mg protein, p < 0.01) and decreased glutathione peroxidase (0.620 vs. 0.910 U/mg protein, p < 0.01) levels as well as catalase (0.463 vs. 4.948 BU/mg protein, p < 0.001) activities were detected; while the plasma and erythrocyte lipid peroxidation and the reduced glutathione (GSH) levels did not change. The results of the investigated prooxidant and the antioxidant status provide evidence that there exists an oxidative stress in children with infantile autism. While investigating the hemorheological parameters of 25 infantile autistic patients, some characteristic pathological parameters were detected: the initial filtration rate (Fi) (0.72 vs. 0.75 p < 0.01) and the clogging rate (CR) (1.926 vs. 2.912, p < 0.01) values of red blood cells (RBC) decreased while the mean transit time (Tc) (8.93 vs. 7.39, p < 0.001) increased suggesting reduced RBC deformability.
Sigma-Aldrich
Catalase from Corynebacterium glutamicum, solution, deep brown, ≥500000 U/mL
Sigma-Aldrich
Catalase from Aspergillus niger, ammonium sulfate suspension, ≥4,000 units/mg protein
Sigma-Aldrich
Glutathione Peroxidase from bovine erythrocytes, lyophilized powder, ≥300 units/mg protein
Sigma-Aldrich
Catalase from bovine liver, aqueous suspension, 40,000-60,000 units/mg protein (E1%/405)
Sigma-Aldrich
Catalase from bovine liver, powder, suitable for cell culture, 2,000-5,000 units/mg protein
Sigma-Aldrich
Catalase from bovine liver, aqueous solution, ≥30,000 units/mg protein
Sigma-Aldrich
Catalase from bovine liver, lyophilized powder, 2,000-5,000 units/mg protein
Sigma-Aldrich
Catalase from bovine liver, aqueous suspension, 10,000-40,000 units/mg protein
Sigma-Aldrich
Catalase from human erythrocytes, ≥90% (SDS-PAGE), buffered aqueous solution, ≥30,000 units/mg protein
Sigma-Aldrich
Catalase from bovine liver, lyophilized powder, ≥10,000 units/mg protein