- Cerebral calcification, osteopetrosis and renal tubular acidosis: is it carbonic anhydrase-II deficiency?
Cerebral calcification, osteopetrosis and renal tubular acidosis: is it carbonic anhydrase-II deficiency?
Saudi journal of kidney diseases and transplantation : an official publication of the Saudi Center for Organ Transplantation, Saudi Arabia (2013-05-04)
Ala A Sh Ali, Sarmad A Al-Mashta
PMID23640632
ABSTRACT
Carbonic anhydrase-II deficiency is an autosomal recessive disorder with a triad of cerebral calcification, osteopetrosis and renal tubular acidosis (often combined proximal and distal). Mental retardation, growth failure, complications of osteopetrosis and other features were all recorded in this syndrome. We present a case of an Iraqi male with all these features and a positive family history.
Sigma-Aldrich
Carbonic Anhydrase from bovine erythrocytes, ≥95% (SDS-PAGE), specific activity ≥3,500 W-A units/mg protein, lyophilized powder
Sigma-Aldrich
Carbonic Anhydrase from bovine erythrocytes, lyophilized powder, ≥2,000 W-A units/mg protein
Sigma-Aldrich
Carbonic Anhydrase Isozyme II from bovine erythrocytes, lyophilized powder, ≥3,000 W-A units/mg protein
Sigma-Aldrich
Carbonic Anhydrase Isozyme II from bovine erythrocytes, lyophilized powder, ≥2,000 W-A units/mg protein
Sigma-Aldrich
Carbonic Anhydrase Isozyme II from bovine erythrocytes, Isoelectric focusing marker, pI 5.4
Sigma-Aldrich
Carbonic Anhydrase II bovine, ≥90% (SDS-PAGE), recombinant, expressed in E. coli, ≥5,000 units/mg protein, buffered aqueous solution
Sigma-Aldrich
Carbonic Anhydrase II human, recombinant, expressed in E. coli, buffered aqueous solution