Merck
  • Gene expression profiling identifies ARSD as a new marker of disease progression and the sphingolipid metabolism as a potential novel metabolism in chronic lymphocytic leukemia.
Gene expression profiling identifies ARSD as a new marker of disease progression and the sphingolipid metabolism as a potential novel metabolism in chronic lymphocytic leukemia.
Cancer biomarkers : section A of Disease markers (2011-01-01)
Alessandra Trojani, Barbara Di Camillo, Alessandra Tedeschi, Milena Lodola, Simona Montesano, Francesca Ricci, Eleonora Vismara, Antonino Greco, Silvio Veronese, Aldo Orlacchio, Sabata Martino, Chiara Colombo, Mariangela Mura, Michele Nichelatti, Anna Colosimo, Barbara Scarpati, Marco Montillo, Enrica Morra
ABSTRACT
Several studies demonstrated IGVH mutational status and ZAP70 expression as the most relevant prognostic markers in Chronic Lymphocytic Leukemia (CLL), suggesting the separation of two patient subgroups: with good mutated ZAP70 negative (MTZAP70(-) and poor unmutated ZAP70 positive (UMZAP70(+)) prognosis. We determined the gene expression of B cells in 112 CLL patients divided into three classes: class 1 with MTZAP70(-), class 2 with UMZAP70(+), and class 3 included both UMZAP70(-) and MTZAP70(+). We found LPL, AGPAT2, MBOAT1, CHPT1, AGPAT4, PLD1 genes encoding enzymes involved in lipid metabolism overexpressed in UMZAP70(+). In addition, this study identified ARSD, a gene belonging to the sphingolipid metabolism, as a new gene significantly overexpressed in UMZAP70(+) compared to MTZAP70(-). Western blots confirmed that ARSD protein levels were significantly different between the 3 classes of patients and normal controls. Statistical analysis identified a significant correlation between ARSD and IGVH; however, both ARSD protein level and IGVH were independently associated with the need for therapy of CLL patients. ARSD is a novel prognostic factor as the time to start therapy is shorter in patients with high levels of ARSD protein and sphingolipid metabolism could represent a new biological mechanism in CLL.
Sigma-Aldrich
Sulfatase from Aerobacter aerogenes, Type VI, buffered aqueous glycerol solution, 2-5 units/mg protein (biuret), 10-20 units/mL
Sigma-Aldrich
Sulfatase from Helix pomatia, Type H-2, aqueous solution, ≥2,000 units/mL
Sigma-Aldrich
Sulfatase from Helix pomatia, Type H-1, sulfatase ≥10,000 units/g solid
Sigma-Aldrich
Sulfatase from Patella vulgata (keyhole limpet), Type IV, essentially salt-free, lyophilized powder, ≥10 units/mg solid
Sigma-Aldrich
Sulfatase from abalone entrails, Type VIII, lyophilized powder, 20-40 units/mg solid