Merck
  • Structural characterization of the N-linked oligosaccharides in bile salt-stimulated lipase originated from human breast milk.
Structural characterization of the N-linked oligosaccharides in bile salt-stimulated lipase originated from human breast milk.
Glycobiology (1999-02-20)
Y Mechref, P Chen, M V Novotny
ABSTRACT
The detailed structures of N- glycans derived from bile salt-stimulated lipase (BSSL) found in human milk were determined by combining exoglycosidase digestion with matrix-assisted laser desorption/ionization time-of-flight mass spectrometry. The N- glycan structures were conclusively determined in terms of complexity and degree of fucosylation. Ion-exchange chromatography with pulsed amperometric detection, together with mass-spectral analysis of the esterified N- glycans, indicated the presence of monosialylated structures. The molecular mass profile of esterified N- glycans present in BSSL further permitted the more detailed studies through collision-induced dissociation (CID) and sequential exoglycosidase cleavages. The N- glycan structures were elucidated to be complex/dibranched, fucosylated/complex/dibranched, monosialylated/complex/dibranched, and monosialylated/fucosylated/dibranched entities.
Sigma-Aldrich
Lipase from Candida sp., recombinant, expressed in Aspergillus niger
Sigma-Aldrich
Lipase from Aspergillus oryzae, lyophilized, powder, white, ~50 U/mg
Sigma-Aldrich
Lipase from Mucor miehei, powder, slightly brown, ~1 U/mg
Sigma-Aldrich
Lipase from Rhizopus oryzae, powder (fine), ~10 U/mg
Sigma-Aldrich
Lipase from Rhizopus niveus, powder (fine), ≥1.5 U/mg
Sigma-Aldrich
Lipase from Aspergillus oryzae, ≥20,000 U/g
Sigma-Aldrich
Lipase acrylic resin, ≥5,000 U/g, recombinant, expressed in Aspergillus niger
Sigma-Aldrich
Lipase from Mucor miehei, lyophilized powder, ≥4,000 units/mg solid (using olive oil)
Sigma-Aldrich
Lipase from Pseudomonas sp., Type XIII, lyophilized powder, ≥15 units/mg solid
Sigma-Aldrich
Lipase from Candida rugosa, lyophilized powder, ≥40,000 units/mg protein
Sigma-Aldrich
Lipase from porcine pancreas, Type II, ≥125 units/mg protein (using olive oil (30 min incubation)), 30-90 units/mg protein (using triacetin)
Sigma-Aldrich
Lipase from porcine pancreas, Type VI-S, ≥20,000 units/mg protein, lyophilized powder
Sigma-Aldrich
Lipase from wheat germ, Type I, lyophilized powder, 5-15 units/mg solid
Sigma-Aldrich
Lipase from Candida rugosa, Type VII, ≥700 unit/mg solid
Sigma-Aldrich
Lipase from Candida rugosa, powder, yellow-brown, ≥2 U/mg
Sigma-Aldrich
Lipase from Candida rugosa, lyophilized, powder (fine), 15-25 U/mg
Sigma-Aldrich
Lipase A Candida antarctica, recombinant from Aspergillus oryzae, powder, beige, ~2 U/mg
Sigma-Aldrich
Lipase B Candida antarctica, recombinant from Aspergillus oryzae, powder, beige, ~9 U/mg
Sigma-Aldrich
Lipase from Pseudomonas cepacia, powder, light beige, ≥30 U/mg
Sigma-Aldrich
Lipase immobilized from Candida antarctica, beads, slightly brown, >2 U/mg
Sigma-Aldrich
Lipase from Mucor javanicus, lyophilized powder, ≥300 units/mg solid (using olive oil)
Sigma-Aldrich
Lipase from Aspergillus niger, powder (fine), ~200 U/g