Merck
A novel endothelial-derived lipase that modulates HDL metabolism.
Nature genetics (1999-04-07)
M Jaye, K J Lynch, J Krawiec, D Marchadier, C Maugeais, K Doan, V South, D Amin, M Perrone, D J Rader
ABSTRACT
High-density lipoprotein (HDL) cholesterol levels are inversely associated with risk of atherosclerotic cardiovascular disease. At least 50% of the variation in HDL cholesterol levels is genetically determined, but the genes responsible for variation in HDL levels have not been fully elucidated. Lipoprotein lipase (LPL) and hepatic lipase (HL), two members of the triacylglyerol (TG) lipase family, both influence HDL metabolism and the HL (LIPC) locus has been associated with variation in HDL cholesterol levels in humans. We describe here the cloning and in vivo functional analysis of a new member of the TG lipase family. In contrast to other family members, this new lipase is synthesized by endothelial cells in vitro and thus has been termed endothelial lipase (encoded by the LIPG gene). EL is expressed in vivo in organs including liver, lung, kidney and placenta, but not in skeletal muscle. In contrast to LPL and HL, EL has a lid of only 19 residues. EL has substantial phospholipase activity, but less triglyceride lipase activity. Overexpression of EL in mice reduced plasma concentrations of HDL cholesterol and its major protein apolipoprotein A-I. The endothelial expression, enzymatic profile and in vivo effects of EL suggest that it may have a role in lipoprotein metabolism and vascular biology.
Sigma-Aldrich
Lipase from Candida sp., recombinant, expressed in Aspergillus niger
Sigma-Aldrich
Lipase from Aspergillus oryzae, lyophilized, powder, white, ~50 U/mg
Sigma-Aldrich
Lipase from Mucor miehei, powder, slightly brown, ~1 U/mg
Sigma-Aldrich
Lipase from Rhizopus oryzae, powder (fine), ~10 U/mg
Sigma-Aldrich
Lipase from Rhizopus niveus, powder (fine), ≥1.5 U/mg
Sigma-Aldrich
Lipase from Aspergillus oryzae, ≥20,000 U/g
Sigma-Aldrich
Lipase acrylic resin, ≥5,000 U/g, recombinant, expressed in Aspergillus niger
Sigma-Aldrich
Lipase from Mucor miehei, lyophilized powder, ≥4,000 units/mg solid (using olive oil)
Sigma-Aldrich
Lipase from Pseudomonas sp., Type XIII, lyophilized powder, ≥15 units/mg solid
Sigma-Aldrich
Lipase from Candida rugosa, lyophilized powder, ≥40,000 units/mg protein
Sigma-Aldrich
Lipase from porcine pancreas, Type II, ≥125 units/mg protein (using olive oil (30 min incubation)), 30-90 units/mg protein (using triacetin)
Sigma-Aldrich
Lipase from porcine pancreas, Type VI-S, ≥20,000 units/mg protein, lyophilized powder
Sigma-Aldrich
Lipase from wheat germ, Type I, lyophilized powder, 5-15 units/mg solid
Sigma-Aldrich
Lipase from Candida rugosa, Type VII, ≥700 unit/mg solid
Sigma-Aldrich
Lipase from Candida rugosa, powder, yellow-brown, ≥2 U/mg
Sigma-Aldrich
Lipase from Candida rugosa, lyophilized, powder (fine), 15-25 U/mg
Sigma-Aldrich
Lipase A Candida antarctica, recombinant from Aspergillus oryzae, powder, beige, ~2 U/mg
Sigma-Aldrich
Lipase B Candida antarctica, recombinant from Aspergillus oryzae, powder, beige, ~9 U/mg
Sigma-Aldrich
Lipase from Pseudomonas cepacia, powder, light beige, ≥30 U/mg
Sigma-Aldrich
Lipase immobilized from Candida antarctica, beads, slightly brown, >2 U/mg
Sigma-Aldrich
Lipase from Mucor javanicus, lyophilized powder, ≥300 units/mg solid (using olive oil)
Sigma-Aldrich
Lipase from Aspergillus niger, powder (fine), ~200 U/g