Merck
  • Identification and characterization of human DNA polymerase beta 2, a DNA polymerase beta -related enzyme.
Identification and characterization of human DNA polymerase beta 2, a DNA polymerase beta -related enzyme.
The Journal of biological chemistry (2000-07-11)
K Nagasawa, K Kitamura, A Yasui, Y Nimura, K Ikeda, M Hirai, A Matsukage, M Nakanishi
ABSTRACT
The BRCA1 COOH terminus (BRCT) motif is present in many nuclear proteins that contribute to cell cycle regulation or DNA repair. Polymerase chain reaction-based screening with degenerate primers targeted to the BRCT motif resulted in the isolation of a human cDNA for a previously unidentified DNA polymerase (designated DNA polymerase beta2) that is closely related to DNA polymerase beta (Pol beta). The predicted Pol beta2 protein contains a BRCT motif in its NH(2)-terminal region; its COOH-terminal region exhibits 33% sequence identity to a corresponding region of human Pol beta. The Pol beta2 gene is expressed in a tissue-specific manner, with transcripts being most abundant in testis. A fusion construct comprising Pol beta2 and green fluorescent protein exhibited a predominantly nuclear localization in transfected HeLa cells. Recombinant human Pol beta2 from insect cells exhibited substantial DNA polymerase activity, but it did not possess terminal deoxyribonucleotidyl transferase activity. A truncated Pol beta2 mutant lacking the BRCT motif retained substantial DNA polymerase activity, whereas a mutant Pol beta2 with two alanine point mutations within the DNA polymerase active site did not. These results indicate that Pol beta2 is a Pol beta-related DNA polymerase with a BRCT motif that is dispensable for its polymerase activity.
Sigma-Aldrich
DNA Polymerase I from Escherichia coli lysogenic for NM 964, buffered aqueous glycerol solution
Sigma-Aldrich
Taq DNA Polymerase from Thermus aquaticus, with 10× PCR reaction buffer without MgCl2
Sigma-Aldrich
Taq DNA Polymerase from Thermus aquaticus, with 10× PCR reaction buffer containing MgCl2