- SPLASH (PLA2IID), a novel member of phospholipase A2 family, is associated with lymphotoxin deficiency.
SPLASH (PLA2IID), a novel member of phospholipase A2 family, is associated with lymphotoxin deficiency.
Genes and immunity (2001-02-24)
A N Shakhov, A V Rubtsov, I G Lyakhov, A V Tumanov, S A Nedospasov
PMID11196711
ABSTRACT
Lymphotoxin (LT) deficient mice have profound defects in the splenic microarchitecture associated with defective expression on certain gene products, including chemokines. By using subtraction cloning of splenic cDNA from wild-type and LT alpha or TNF/LT alpha double deficient mice we isolated a novel murine gene encoding a secretory type phospholipase A2, called SPLASH. The two major alternative transcripts of SPLASH gene are predominantly expressed in lymphoid tissues, such as spleen and lymph nodes. SPLASH maps to the distal part of chromosome 4, to which several cancer-related loci have been also mapped.
Sigma-Aldrich
Phospholipase A2 from honey bee venom (Apis mellifera), salt-free, lyophilized powder, 600-2400 units/mg protein
Sigma-Aldrich
Phospholipase A2 from porcine pancreas, ammonium sulfate suspension, ≥600 units/mg protein