Merck
  • System-wide temporal characterization of the proteome and phosphoproteome of human embryonic stem cell differentiation.
System-wide temporal characterization of the proteome and phosphoproteome of human embryonic stem cell differentiation.
Science signaling (2011-03-17)
Kristoffer T G Rigbolt, Tatyana A Prokhorova, Vyacheslav Akimov, Jeanette Henningsen, Pia T Johansen, Irina Kratchmarova, Moustapha Kassem, Matthias Mann, Jesper V Olsen, Blagoy Blagoev
ABSTRACT
To elucidate cellular events underlying the pluripotency of human embryonic stem cells (hESCs), we performed parallel quantitative proteomic and phosphoproteomic analyses of hESCs during differentiation initiated by a diacylglycerol analog or transfer to media that had not been conditioned by feeder cells. We profiled 6521 proteins and 23,522 phosphorylation sites, of which almost 50% displayed dynamic changes in phosphorylation status during 24 hours of differentiation. These data are a resource for studies of the events associated with the maintenance of hESC pluripotency and those accompanying their differentiation. From these data, we identified a core hESC phosphoproteome of sites with similar robust changes in response to the two distinct treatments. These sites exhibited distinct dynamic phosphorylation patterns, which were linked to known or predicted kinases on the basis of the matching sequence motif. In addition to identifying previously unknown phosphorylation sites on factors associated with differentiation, such as kinases and transcription factors, we observed dynamic phosphorylation of DNA methyltransferases (DNMTs). We found a specific interaction of DNMTs during early differentiation with the PAF1 (polymerase-associated factor 1) transcriptional elongation complex, which binds to promoters of the pluripotency and known DNMT target genes encoding OCT4 and NANOG, thereby providing a possible molecular link for the silencing of these genes during differentiation.
Sigma-Aldrich
Lipase from Candida sp., recombinant, expressed in Aspergillus niger
Sigma-Aldrich
Adenosine 5′-Triphosphatase from porcine cerebral cortex, lyophilized powder, ≥0.3 units/mg protein, pH 7.8
Sigma-Aldrich
Sphingomyelinase from Staphylococcus aureus, buffered aqueous glycerol solution, 100-300 units/mg protein (Lowry)
Sigma-Aldrich
Lipase from Mucor miehei, lyophilized powder, ≥4,000 units/mg solid (using olive oil)
Sigma-Aldrich
Phospholipase A2 from honey bee venom (Apis mellifera), salt-free, lyophilized powder, 600-2400 units/mg protein
Sigma-Aldrich
Pyruvate Kinase from rabbit muscle, Type III, lyophilized powder, 350-600 units/mg protein
Sigma-Aldrich
Uridine-5′-diphosphoglucose pyrophosphorylase from baker′s yeast, Type X, lyophilized powder, ≥40 units/mg protein
Sigma-Aldrich
Phosphoglucomutase from rabbit muscle, ammonium sulfate suspension, ≥100 units/mg protein
Sigma-Aldrich
Lipase from Candida rugosa, lyophilized powder, ≥40,000 units/mg protein
Sigma-Aldrich
Lipase from Pseudomonas sp., Type XIII, lyophilized powder, ≥15 units/mg solid
Sigma-Aldrich
Lipase from porcine pancreas, Type II, ≥125 units/mg protein (using olive oil (30 min incubation)), 30-90 units/mg protein (using triacetin)
Sigma-Aldrich
Pyruvate Kinase from rabbit muscle, Type II, ammonium sulfate suspension, 350-600 units/mg protein
Sigma-Aldrich
Lipase from wheat germ, Type I, lyophilized powder, 5-15 units/mg solid
Sigma-Aldrich
Lipase from porcine pancreas, Type VI-S, ≥20,000 units/mg protein, lyophilized powder
Sigma-Aldrich
Taq DNA Polymerase from Thermus aquaticus, with 10× PCR reaction buffer without MgCl2
Sigma-Aldrich
Taq DNA Polymerase from Thermus aquaticus, with 10× PCR reaction buffer containing MgCl2
Sigma-Aldrich
Lipase from Candida rugosa, Type VII, ≥700 unit/mg solid
Supelco
GAPDH, standard for protein electrophoresis
Sigma-Aldrich
Glyceraldehyde-3-phosphate Dehydrogenase from rabbit muscle, lyophilized powder, ≥75 units/mg protein
Sigma-Aldrich
Pyruvate Kinase from rabbit muscle, Type VII, buffered aqueous glycerol solution, 350-600 units/mg protein
Sigma-Aldrich
Lipase from Rhizopus oryzae, powder (fine), ~10 U/mg
Sigma-Aldrich
Lipase from Aspergillus oryzae, lyophilized, powder, white, ~50 U/mg
Sigma-Aldrich
Lipase from Aspergillus oryzae, ≥20,000 U/g
Sigma-Aldrich
Lipase from Rhizopus niveus, powder (fine), ≥1.5 U/mg
Sigma-Aldrich
Lipase from Candida rugosa, powder, yellow-brown, ≥2 U/mg
Sigma-Aldrich
Lipase acrylic resin, ≥5,000 U/g, recombinant, expressed in Aspergillus niger
Sigma-Aldrich
Lipase from Candida rugosa, lyophilized, powder (fine), 15-25 U/mg
Sigma-Aldrich
Lipase from Mucor miehei, powder, slightly brown, ~1 U/mg
Sigma-Aldrich
Acetyl-CoA carboxylase 2 human, recombinant, expressed in Sf9 cells
Sigma-Aldrich
Triosephosphate Isomerase from baker′s yeast (S. cerevisiae), Type I, ammonium sulfate suspension, ~10,000 units/mg protein