Merck
  • 5-HT2-receptor antagonists: α1- vs. 5-HT2-receptor blocking propzerties in blood vessels.
5-HT<SUB>2</SUB>-receptor antagonists: &#945;<SUB>1</SUB>- vs. 5-HT<SUB>2</SUB>-receptor blocking propzerties in blood vessels.
Journal of Cardiovascular Pharmacology (1988)
Cohen, et al.