Merck
  • Simultaneous determination of dimethylarsinic acid and monomethylarsonic acid after derivatization with thioglycol methylate by gas chromatography/mass spectrometry.
Simultaneous determination of dimethylarsinic acid and monomethylarsonic acid after derivatization with thioglycol methylate by gas chromatography/mass spectrometry.
Rapid communications in mass spectrometry : RCM (1999-04-21)
Z Mester, G Horváth, G Vitányi, L Lelik, P Fodor
ABSTRACT
A gas chromatography/mass spectrometry (GC/MS) method has been developed to determine two methylated arsenic species in human urine samples. The yield of derivatization for dimethylarsinic acid (DMA) and monomethylarsonic acid (MMA) using thioglycol methylate (TGM) was measured. The detection limit for the derivatized DMA and MMA using the GC/MS method are 0.95 and 0.8 ng cm-3, respectively. This simple and rapid method has good precision and accuracy. Fragmentation routes of derivatized MMA and DMA are suggested on accurate mass measurements.
Sigma-Aldrich
Methyl thioglycolate, 95%