Merck
  • Conformational changes at the active site of pantetheine hydrolase during denaturation by guanidine hydrochloride.
Conformational changes at the active site of pantetheine hydrolase during denaturation by guanidine hydrochloride.
Journal of protein chemistry (2000-02-26)
G Pitari, A A D'Archivio, L Di Leandro, G Antonini, A Panatta, E Tettamanti, S Duprè, F Malatesta
ABSTRACT
Conformational changes at the active site of pantetheine hydrolase (EC3.5.1.-) during guanidine hydrochloride (GndHCl) denaturation were investigated by UV and circular dichroism spectroscopy and by electron spin resonance spectroscopy, following the spectral behaviour of the nitroxide radicals (N-(1-oxyl-2,2,5,5,-tetramethyl-3-pyrrolidinyl) iodacetamide) covalently linked to the two active site cysteine residues. At low denaturant concentrations (0.2 M) no conformational changes may be observed, whereas the catalytic activity, is strongly affected. The results indicate that the active site of pantetheine hydrolase is labile and unfolds under conditions in which no global tertiary structure modifications can be observed.