Merck
  • Spectrophotometric determination of fluoxetine hydrochloride in bulk and in pharmaceutical formulations.
Spectrophotometric determination of fluoxetine hydrochloride in bulk and in pharmaceutical formulations.
Journal of pharmaceutical and biomedical analysis (2000-03-04)
A H Prabhakar, V B Patel, R Giridhar
ABSTRACT
Two new rapid, sensitive and economical spectrophotometric methods are described for the determination of fluoxetine hydrochloride in bulk and in pharmaceutical formulations. Both methods are based on the formation of a yellow ion-pair complex due to the action of methyl orange (MO) and thymol blue (TM) on fluoxetine in acidic (pH 4.0) and basic (pH 8.0) medium, respectively. Under optimised conditions they show an absorption maxima at 433 nm (MO) and 410 nm (TB), with molar absorptivities of 2.12 x 10(-4) and 4.207 x 10(-3) l mol(-1) cm(-1) and Sandell's Sensitivities of 1.64 x 10(-2) and 0.082 microg cm(-2) per 0.001 absorbance unit for MO and TB, respectively. The colour is stable for 5 min after extraction. In both cases Beer's Law is obeyed at 1-20 microg mol(-1) with MO and 4-24 microg mol(-1) with TB. The proposal method was successfully extended to pharmaceutical preparations capsules. The results obtained by both the agreement and E.P. (3rd edition) were in good agreement and statistical comparison by Student's t-test and variance ratio F-test showed no significant difference in the three methods.
Product Description
Supelco
Thymolphthalein, ACS reagent
Sigma-Aldrich
Thymolphthalein, ACS reagent, Dye content 95 %
Sigma-Aldrich
Thymol Blue, ACS reagent
Sigma-Aldrich
Thymol Blue sodium salt, ACS reagent, Dye content 95 %