Merck
  • Fast Green FCF (Food Green 3) inhibits synaptic activity in rat hippocampal interneurons.
Fast Green FCF (Food Green 3) inhibits synaptic activity in rat hippocampal interneurons.
Neuroscience letters (2002-01-23)
Johannes A van Hooft
ABSTRACT
The effects of the certified food dye Fast Green FCF (Food Green 3) on miniature synaptic events in whole-cell voltage clamped hippocampal interneurons were examined. Fast Green FCF reduced the frequency, but did not affect the amplitude or kinetics, of miniature synaptic events in a dose-dependent manner. At 1 mg/ml (1.2 mM), the frequency was reduced to 21% of control. The half-maximum effective concentration was estimated to be 200 microg/ml (250 microM). In contrast, the dye Phenol Red neither affected frequency, amplitude nor kinetics of synaptic events. The results suggest that Fast Green FCF acts at a presynaptic locus, thereby inhibiting the release of neurotransmitter.
Product Description
Supelco
Fast Green FCF, analytical standard
Sigma-Aldrich
Fast Green FCF, certified by the BSC
Sigma-Aldrich
Fast Green FCF, Dye content ≥85 %