Merck
  • Potentiation and prolongation of the insulinotropic action of glucagon-like peptide 1 by methyl pyruvate or dimethyl ester of L-glutamic acid in a type 2 diabetes animal model.
Potentiation and prolongation of the insulinotropic action of glucagon-like peptide 1 by methyl pyruvate or dimethyl ester of L-glutamic acid in a type 2 diabetes animal model.
Endocrine (2002-03-13)
J Cancelas, M L Villanueva-Peñacarrillo, I Valverde, W J Malaisse
ABSTRACT
Methyl pyruvate and the dimethyl ester of L-glutamic acid were administered intravenously, as a primed constant infusion (1.0-2.0 micromol followed by 0.5-1.0 micromol/min, both expressed per gram of body wt), in adult rats that had been injected with streptozotocin during the neonatal period. Each ester augmented plasma insulin concentration and potentiated and/or prolonged the insulinotropic action of glucagon-like peptide 1 (GLP-1) injected intravenously (5 pmol/g of body wt) at min 5 of the test. It is proposed, therefore, that suitable nonglucidic nutrients, susceptible to bypassing the site-specific defects of D-glucose transport and metabolism responsible for the preferential impairment of the B-cell secretory response to D-glucose in non-insulin-dependent diabetes, could be used to optimize the insulinotropic action of GLP-1.
Sigma-Aldrich
L-Glutamic acid dimethyl ester hydrochloride, ≥99.0% (anhydrous basis material, AT)
Sigma-Aldrich
Methyl pyruvate, 90%, technical grade