Merck
  • Shock structures and bunching fronts in excitable reaction-diffusion systems.
Shock structures and bunching fronts in excitable reaction-diffusion systems.
Physical review. E, Statistical, nonlinear, and soft matter physics (2002-05-15)
Chad T Hamik, Oliver Steinbock
ABSTRACT
We report experimental results on the dynamics of excitation waves in a modified Belousov-Zhabotinsky reaction. The waves in this system obey nonmonotonic dispersion relations. This anomaly induces the stacking of excitation fronts into patterns with stable interpulse distances. The stacking process creates either a traveling shock structure or a cascade of bunching events in which metastable wave packets are formed. The direction and the speed of the shock are explained in terms of a simple geometrical analysis. We also present experimental evidence for the corresponding instabilities in two-dimensional systems. Here, wave stacking generates atypical structures in the collision of target patterns and wave bunching is accompanied by complex front deformations.
Sigma-Aldrich
Ferroin indicator solution, 0.1 wt. % in H2O