Merck
  • The trouble with teething--misdiagnosis and misuse of a topical medicament.
The trouble with teething--misdiagnosis and misuse of a topical medicament.
International journal of paediatric dentistry (2002-05-25)
P H R Wilson, C Mason
ABSTRACT
For many clinicians and parents "teething" remains a convenient diagnosis to explain all manner of local and systemic upset in the young child. Many therapies are on the market to help alleviate the symptoms of primary tooth eruption. In this article we highlight the problems of "teething" as a diagnosis by presenting a case where an initial misdiagnosis of teething compromised a patient's life. The same patient then suffered from topical analgesic misuse during the recovery period.
Sigma-Aldrich
Benzyldimethylhexadecylammonium chloride, cationic detergent