Merck
  • Synthesis of (+/-)-5-epi-citreoviral and (+/-)-citreoviral and the kinetic resolution of an allylic silane by a [3 + 2] annulation.
Synthesis of (+/-)-5-epi-citreoviral and (+/-)-citreoviral and the kinetic resolution of an allylic silane by a [3 + 2] annulation.
Organic letters (2002-08-17)
Zhi-Hui Peng, K A Woerpel
ABSTRACT
[reaction: see text] The [3 + 2] annulation reaction of allylsilane 1 with an alpha-keto ester provided the highly substituted tetrahydrofuran 2 as a single diastereomer in high yield. The synthesis of (+/-)-5-epi-citreoviral and (+/-)-citreoviral has been accomplished with this annulation reaction as the key step. Using the pantolactone-derived alpha-keto ester, the allylsilane 1 has been resolved with high enantiomeric purity.
Sigma-Aldrich
D-(−)-Pantolactone, 99%
Sigma-Aldrich
(S)-(+)-Pantolactone, 97%
Sigma-Aldrich
DL-α-Hydroxy-β,β-dimethyl-γ-butyrolactone, purum, ≥97.0% (T)