Merck
  • New substrates for enkephalinase (neutral endopeptidase) based on fluorescence energy transfer.
New substrates for enkephalinase (neutral endopeptidase) based on fluorescence energy transfer.
Biochemical and biophysical research communications (1987-02-27)
B Malfroy, J Burnier
ABSTRACT
Novel fluorescent substrates for enkephalinase (neutral endopeptidase; EC 3.4.24.11) have been developed. These new assays are based on the disappearance of energy transfer between a tryptophan or a tyrosine residue and the 5-dimethylaminonaphthalene-1-sulfonyl group (dansyl) in the substrates dansyl-Gly-Trp-Gly or dansyl-Gly-Tyr-Gly upon hydrolysis of their Gly-Trp or Gly-Tyr amide bond by enkephalinase. No significant difference in Km or kcat values were found for dansyl-Gly-Trp-Gly and dansyl-Gly-Tyr-Gly, indicating that, in contrast to thermolysin, the active site of enkephalinase easily accommodates tryptophan residues. Both tryptophan and tyrosine-containing substrates can be used for continuous recording of enkephalinase activity and should prove useful for detailed study of the substrate specificity of this enzyme.