Merck
  • High susceptibility of p53 knockout mice to esophageal and urinary bladder carcinogenesis induced by N, N-dibutylnitrosamine.
High susceptibility of p53 knockout mice to esophageal and urinary bladder carcinogenesis induced by N, N-dibutylnitrosamine.
Cancer letters (2003-04-23)
Takayuki Nishikawa, Elsayed I Salim, Keiichirou Morimura, Masahiro Kaneko, Motome Ogawa, Anna Kinoshita, Harushi Osugi, Hiroaki Kinoshita, Shoji Fukushima
ABSTRACT
In human cancer, alterations in the p53 tumor suppressor gene are the most common genetic alterations. The aim of the present study was to detect sensitivity of the p53 (+/-) mice and their littermates p53 (+/+) mice to N, N-dibutylnitrosamine (DBN) carcinogenicity. In experiment 1, 6-7-week-old p53 (+/-) and p53 (+/+) mice were treated with 0, 0.025 and 0.05% DBN, respectively, in drinking water for 20 weeks. Esophageal squamous cell and urinary bladder transitional cell carcinomas (TCCs) and fibrosarcomas were found to be significantly increased in p53 (+/-) mice treated with doses of DBN compared to p53 (+/+) mice administered similar doses. In experiment 2, 6-7-week-old p53 (+/-) and p53 (+/+) mice were administered 0 or 0.05 % DBN in drinking water for 8 weeks. Immunohistochemical staining revealed a significant increase in numbers of p53 and bromodeoxyuridine (BrdU) positive cells in the esophageal and urinary bladder epithelia of DBN-treated p53 (+/-) mice compared to p53 (+/+) mice administered DBN. Molecular analysis revealed point mutations in the residual p53 allele in four of eight (50%) esophageal mucosa of DBN-treated p53 (+/-) mice, and in three of eight (38%) of treated p53 (+/+) mice. The results show that p53 (+/-) mice were sensitive to DBN treatment with respect to esophageal and bladder tumor development, with a mechanism that could be confined to early mutations of the residual p53 allele and increased cellular proliferation in the target organs.
Product Description
Supelco
N-Nitrosodi-n-butylamine, analytical standard
Supelco
N-Nitrosodi-n-butylamine solution, certified reference material, 2000 μg/mL in methylene chloride