- Cyclopentanes from N-amino-glyconolactams. A synthesis of mannostatin A.
Cyclopentanes from N-amino-glyconolactams. A synthesis of mannostatin A.
Chemical communications (Cambridge, England) (2003-05-15)
Guixian Hu, Martin Zimmermann, Chepuri Venkata Ramana, Andrea Vasella
PMID12744315
ABSTRACT
Oxidation of a N-amino-ribonolactam with lead tetraacetate yields two cyclopentanes; the major one was transformed into the alpha-mannosidase inhibitor mannostatin A.