Merck
  • Stannyl radical addition-cyclization of oxime ethers connected with olefins.
Stannyl radical addition-cyclization of oxime ethers connected with olefins.
Chemical & pharmaceutical bulletin (2004-01-08)
Hideto Miyabe, Hirotaka Tanaka, Takeaki Naito
ABSTRACT
Stannyl radical addition-cyclization of oxime ethers connected with olefin moieties was studied. The radical reactions proceeded effectively by the use of triethylborane as a radical initiator to provide the functionalized pyrrolidines via a carbon-carbon bond-forming process.
Sigma-Aldrich
Triethylborane solution, 1.0 M in hexanes
Sigma-Aldrich
Triethylborane solution, 1.0 M in THF