Merck
  • In vitro effects of the dicyclohexylammonium salt of hyperforin on interleukin-6 release in different experimental models.
In vitro effects of the dicyclohexylammonium salt of hyperforin on interleukin-6 release in different experimental models.
Planta medica (2004-08-12)
Marco Gobbi, Manuela Moia, Marcella Funicello, Antonella Riva, Paolo Morazzoni, Tiziana Mennini
ABSTRACT
Cytokine hypersecretion might be involved in the onset and maintenance of depressive disorders and it has been suggested that St. John's wort extracts (Hypericum perforatum, SJW) might exert their antidepressant-like effects by affecting peripheral interleukin-6 (IL6) expression. We found that hyperforin, one putative active principle of SJW, and its dicyclohexylammonium salt (hyperforin-DCHA), inhibited the substance P (SP)-induced [L6 release inhuman astrocytoma cells (U373MG) with an Cs50 of 1.6 pM, indicating that hyperforin is likely to account for the inhibitory effect previously found in the same experimental model with SJW ex-tracts. [3H]SP binding experiments in parallel on the same intact cells indicate that hyperforin-DCHA does not interact with neuro-kinin-I receptors but very likely interacts with some intracellular steps leading to the synthesis and/or release of IL6. Hyperforin-DCHA also inhibited, with a similar IC50, the IL6 release induced in U373MG cells by two other classic proinflammatory stimuli,ILl and lipopolysaccharide (LPS), as well as the LPS-induced IL6 release in whole rat blood. Hyperforin-DCHA was less active in whole human blood. The concentrations required in vitro to inhibit LPS-induced IL6 release from rat and human whole blood are about one order of magnitude higher than the hyperforin levels measured in the plasma of rats or humans treated with pharmaco-logically active doses of SJW or hyperforin-DCHA.
Sigma-Aldrich
Dicyclohexylamine, 99%
Sigma-Aldrich
Dicyclohexylamine nitrite, 97%