Merck
Plasticizer metabolites in the environment.
Water research (2004-09-08)
Owen Horn, Sandro Nalli, David Cooper, Jim Nicell
ABSTRACT
Earlier work with pure cultures had shown that the interaction of microbes with plasticizers leads to the formation of metabolites including 2-ethylhexanoic acid and 2-ethylhexanol that resist further degradation. The presence of these metabolites is now reported in a variety of environmental samples. Thus, even in a complex ecosystem, when plasticizers are degraded, the breakdown is not complete and significant amounts of 2-ethylhexanoic acid and 2-ethylhexanol are observed. These compounds have been shown to exhibit acute toxicity using Microtox, Daphnia, rainbow trout and fathead minnow toxicity assays. Since it is already well established that plasticizers are ubiquitous in the environment, it is expected that their recalcitrant metabolites will also be ubiquitous. This is of concern because, while the plasticizers do not exhibit acute toxicity, their metabolites do.
Sigma-Aldrich
2-Ethyl-1-hexanol, ≥99%, FG
Sigma-Aldrich
2-Ethyl-1-hexanol, ≥99.6%
Supelco
2-Ethyl-1-hexanol, analytical standard
Sigma-Aldrich
2-Ethyl-1-hexanol, puriss., ≥99.0% (GC)
Sigma-Aldrich
Titanium(IV) 2-ethylhexyloxide, 95%
Sigma-Aldrich
2-Ethylhexanoic acid, ≥99%
Sigma-Aldrich
2-Ethylhexanoic acid, ≥99%
Sigma-Aldrich
2-Ethylhexanoic acid, 99%
Supelco
2-Ethylhexanoic acid, analytical standard