Merck
  • First successful application of diphosphite ligands in the asymmetric hydroformylation of dihydrofurans.
First successful application of diphosphite ligands in the asymmetric hydroformylation of dihydrofurans.
Chemical communications (Cambridge, England) (2005-02-24)
Montserrat Dieguez, Oscar Pamies, Carmen Claver
ABSTRACT
Good enantioselectivities and excellent regioselectivities are achieved in the Rh-catalyzed asymmetric hydroformylation of 2,5- and 2,3-dihydrofuran using diphosphite ligands; whereby the backbone of the ligand is crucial to suppressing isomerization and obtaining high ee's.
Sigma-Aldrich
2,3-Dihydrofuran, 99%