Merck
  • Engaging the pyridine-DMAD zwitterion in a novel strategy for the selective synthesis of highly substituted benzene and cyclopentenedione derivatives.
Engaging the pyridine-DMAD zwitterion in a novel strategy for the selective synthesis of highly substituted benzene and cyclopentenedione derivatives.
Organic letters (2005-10-08)
Vijay Nair, Abhilash N Pillai, P B Beneesh, Eringathodi Suresh
ABSTRACT
[reaction: see text] The pyridine-mediated reaction of dimethyl acetylenedicarboxylate and cyclobutene-1,2-diones affords selective access to either hexasubstituted benzene derivatives or cyclopentenedione derivatives depending on the concentration of pyridine.
Sigma-Aldrich
Dimethyl acetylenedicarboxylate, 95%