Merck
  • New application of air-drying techniques for studying Ephemeroptera and Plecoptera eggs by scanning electron microscopy.
New application of air-drying techniques for studying Ephemeroptera and Plecoptera eggs by scanning electron microscopy.
Microscopy research and technique (2005-11-30)
Nicolás Ubero-Pascal, José Manuel Fortuño, María de Los Angeles Puig
ABSTRACT
Hexamethyldisilizane (HMDS) and tetramethylsilane are organic compounds that are volatile at ambient temperature and which can therefore be used for air-drying biological samples for SEM studies. The techniques using these compounds provide results that are comparable with those obtained by critical point drying, but which involve a very simple process that saves time and money. Both techniques were applied to SEM studies of Ephemeroptera and Plecoptera eggs in order to assess their suitability as alternative methods to critical point drying for these kinds of biological material. The results show no morphological differences between eggs HMDS air-dried and critical point-dried.
Sigma-Aldrich
Tetramethylsilane, electronic grade, ≥99.99% trace metals basis
Supelco
Tetramethylsilane, analytical standard, suitable for NMR (spectroscopy), ACS reagent
Sigma-Aldrich
Tetramethylsilane, ≥99.0% (GC)
Sigma-Aldrich
Tetramethylsilane, NMR grade, ACS reagent, ≥99.9%