Merck
  • Radical reaction of sodium hypophosphite with terminal alkynes: synthesis of 1,1-bis-H-phosphinates.
Radical reaction of sodium hypophosphite with terminal alkynes: synthesis of 1,1-bis-H-phosphinates.
Organic letters (2005-12-16)
Sonia Gouault-Bironneau, Sylvine Deprèle, Amber Sutor, Jean-Luc Montchamp
ABSTRACT
[reaction: see text] The room-temperature radical addition of sodium hypophosphite to terminal alkynes produces the previously unknown 1-alkyl-1,1-bis-H-phosphinates in moderate yield. The reaction is initiated by R3B and air and proceeds under mild conditions in an open container. The bissodium salts precipitate spontaneously from the reaction mixtures, thus providing a simple purification procedure and the opportunity for multigram synthesis. The 1,1-bis-H-phosphinate products are novel precursors of the biologically important 1,1-bisphosphonates.
Sigma-Aldrich
Sodium hypophosphite monohydrate, ≥99%
Sigma-Aldrich
Sodium hypophosphite, puriss., meets analytical specification of BPC63, anhydrous, 98-101%