Merck
  • Amidrazones as precursors of biologically active compounds--synthesis of diaminopyrazoles for evaluation of anticancer activity.
Amidrazones as precursors of biologically active compounds--synthesis of diaminopyrazoles for evaluation of anticancer activity.
Archiv der Pharmazie (2006-01-18)
Maria Teresa Cocco, Cenzo Congiu, Valentina Lilliu, Valentina Onnis
ABSTRACT
The regioselectivity of coupling phenyl isocyanate to 3-(2-acylhydrazino)-3-aminopropenenitriles and ethyl 3-(2-acylhydrazino)-3-aminopropenoates as simple access to aminopyrazole derivatives, endowed with potential antitumoral activity, is reported. 3-(2-Acylhydrazino)-3-aminopropenenitriles react with phenyl isocyanate to afford 3-amino-3-(2-acylhydrazino)-2-phenylaminocarbonyl-2-propenenitriles. These key intermediates were cyclized into 3,5-diaminopyrazole-4-carboxamide derivatives. Preliminary results of poor antiproliferative activities of these compounds are also reported.
Product Description
Supelco
Phenyl isocyanate, derivatization grade (HPLC), LiChropur, ≥99.0% (GC)
Sigma-Aldrich
Phenyl isocyanate, ≥98%