Merck
  • Hair analysis of histamine and several metabolites in C3H/HeNCrj mice by ultra performance liquid chromatography with electrospray ionization time-of-flight mass spectrometry (UPLC-ESI-TOF-MS): influence of hair cycle and age.
Hair analysis of histamine and several metabolites in C3H/HeNCrj mice by ultra performance liquid chromatography with electrospray ionization time-of-flight mass spectrometry (UPLC-ESI-TOF-MS): influence of hair cycle and age.
Clinica chimica acta; international journal of clinical chemistry (2007-01-02)
Hiroki Kawanishi, Toshimasa Toyo'oka, Kenichi Ito, Munenori Maeda, Tomoji Hamada, Takeshi Fukushima, Masaru Kato, Shinsuke Inagaki
ABSTRACT
According to a previous study, the concentration of HA in the hair of SD rats was similar in each rat and the variation in HA concentration was not so great. However, the concentration in human hair was fairly different in each person. As possible reasons for the higher variation in human hair, the differences in hair cycles and age in each person may be considerable. Based on this idea, the studies using C3H/HeNCrj mice who can synchronize their hair cycle were performed for resolution of the influence of hair cycle and age. The effects of hair cycle and age on the concentration of histamine (HA) and several metabolites, i.e., 1-methylhistamine (MHA), imidazole-4-acetic acid (IAA), and 1-methyl-4-imidazole-acetic acid (MIAA), in C3H/HeNCrj mice hair were investigated by ultra-performance liquid chromatography (UPLC) with electrospray ionization time-of-flight mass spectrometry (ESI-TOF-MS). HA and the metabolites were labeled with 4-(N,N-dimethylaminosulfonyl)-7-fluoro-2,1,3-benzoxadiazole (DBD-F) and 4-(N,N-dimethylaminosulfonyl)-7-piperazino-2,1,3-benzoxadiazole (DBD-PZ). The resulting derivatives were separated by UPLC and determined with ESI-TOF-MS. A good linearity was achieved from the calibration curves, obtained by plotting the peak area ratios of the analytes relative to the internal standard (IS), i.e., histamine-alpha,alpha,beta,beta-d4 (HA-d4) or 4-imidazolecarboxylic acid (ICA), against the injected amounts of each compound. The detection limits of HA, MHA, IAA, and MIAA on mass chromatograms were 0.21, 1.0, 0.17, and 0.11 pmol, respectively. The concentrations of HA and the metabolites in the hair shafts and hair root of C3H/HeNCrj mice were determined by this method. The concentration of HA in the hair shaft was relatively higher in the telogen phase. In contrast, the HA content in the anagen phase was increased only in the hair root of old mice. HA appears to possess some effect on hair growth, although the exact reason was not obvious. The HA metabolites, i.e., MHA, MIAA and IAA, were also determined the same as HA; however, the difference in the metabolite concentrations between the hair cycle and age was not clear in both hair shaft and hair root. Such studies of the effect of hair cycle and age on these concentrations are the first report. This analytical technique may be applicable to the determination of various biological compounds in hair.
Sigma-Aldrich
1-Methylhistamine dihydrochloride, ≥98% (TLC), powder
Sigma-Aldrich
4-Imidazolecarboxylic acid, 98%
Sigma-Aldrich
4-Imidazoleacetic acid hydrochloride, 98%