Merck
  • Biotransformation of raspberry ketone and zingerone by cultured cells of Phytolacca americana.
Biotransformation of raspberry ketone and zingerone by cultured cells of Phytolacca americana.
Phytochemistry (2007-01-09)
Kei Shimoda, Toshio Harada, Hatsuyuki Hamada, Nobuyoshi Nakajima, Hiroki Hamada
ABSTRACT
The biotransformation of raspberry ketone and zingerone were individually investigated using cultured cells of Phytolacca americana. In addition to (2S)-4-(4-hydroxyphenyl)-2-butanol (2%), (2S)-4-(3,4-dihydroxyphenyl)-2-butanol (5%), 4-[4-(beta-d-glucopyranosyloxy)phenyl]-2-butanone (19%), 4-[(3S)-3-hydroxybutyl]phenyl-beta-d-glucopyranoside (23%), and (2S)-4-(4-hydroxyphenyl)but-2-yl-beta-d-glucopyranoside (20%), two biotransformation products, i.e., 2-hydroxy-4-[(3S)-3-hydroxybutyl]phenyl-beta-d-glucopyranoside (12%) and 2-hydroxy-5-[(3S)-3-hydroxybutyl]phenyl-beta-d-glucopyranoside (11%), were isolated from suspension cells after incubation with raspberry ketone for three days. On the other hand, two compounds, i.e., (2S)-4-(4-hydroxy-3-methoxyphenyl)but-2-yl-beta-d-glucopyranoside (17%) and (2S)-2-(beta-d-glucopyranosyloxy)-4-[4-(beta-d-glucopyranosyloxy)-3-methoxyphenyl]butane (16%), together with (2S)-4-(4-hydroxy-3-methoxyphenyl)-2-butanol (15%), 4-[4-(beta-d-glucopyranosyloxy)-3-methoxyphenyl]-2-butanone (21%), and 4-[(3S)-3-hydroxybutyl]-2-methoxyphenyl-beta-d-glucopyranoside (24%) were obtained upon addition of zingerone. Cultured cells of P. americana can reduce, and regioselectively hydroxylate and glucosylate, these food ingredients to their beta-glycosides.
Sigma-Aldrich
4-(4-Hydroxyphenyl)-2-butanone, 99%
Sigma-Aldrich
4-(4-Hydroxyphenyl)-2-butanone, ≥98%, FCC, FG
Sigma-Aldrich
4-(4-Hydroxyphenyl)-2-butanone, natural, ≥98%, FCC, FG
Sigma-Aldrich
Vanillylacetone, ≥96%, FG