Merck
  • Identification of urinary metabolites of orally administered N,N-dimethyl-p-toluidine in male F344 rats.
Identification of urinary metabolites of orally administered N,N-dimethyl-p-toluidine in male F344 rats.
Journal of toxicology and environmental health. Part A (2007-04-25)
Nam-Cheol Kim, Katayoon Ghanbari, Dean A Kracko, Waylon M Weber, Jacob D McDonald, Kelly J Dix
ABSTRACT
The metabolism of orally administered N,N-dimethyl-p-toluidine (DMPT) in male F344 rats was investigated. The rat urinary metabolite profile was determined by analytical reverse-phase high performance liquid chromatography (HPLC). Four radiolabeled peaks were observed, isolated, and purified by solid-phase extraction (SPE) and preparative HPLC methods. The 4 peaks were identified as p-(N-acetylhydroxyamino)hippuric acid (M1), DMPT N-oxide (M2), N-methyl-p-toluidine (M3), and parent DMPT. Metabolites M1 and M2 were identified by spectrometric and spectroscopic methods, including mass fragmentation pattern identification from both liquid chromatography/mass spectrometry and gas chromatography/mass spectrometry, and from chemical analysis of nuclear magnetic resonance spectra. Structural confirmation of metabolite M2 was accomplished by comparison with a synthetic standard. Peaks M3 and the peak suspected to be DMPT were identified by comparison of their HPLC retention times and mass fragmentation patterns with authentic standards of N-methyl-p-toluidine and DMPT, respectively. DMPT metabolism is similar to that reported for N,N-dimethylaniline.
Sigma-Aldrich
4,N,N-Trimethylaniline, 99%
Sigma-Aldrich
4,N,N-Trimethylaniline, ≥98.5% (GC)
Sigma-Aldrich
4,N,N-Trimethylaniline, purum, ≥98.0% (GC)