Merck
  • 1,2-Naphthoquinone disrupts the function of cAMP response element-binding protein through covalent modification.
1,2-Naphthoquinone disrupts the function of cAMP response element-binding protein through covalent modification.
Biochemical and biophysical research communications (2007-07-31)
Akiko Endo, Daigo Sumi, Yoshito Kumagai
ABSTRACT
1,2-Naphthoquinone (1,2-NQ) is an atmospheric contaminant with electrophilic properties that allow it to react readily with protein thiol groups such as those found on the cAMP response element-binding protein (CREB), a transcription factor with conserved cysteine residues that regulate DNA binding. In the present study, we explored the possibility that the interaction of 1,2-NQ with CREB will affect its activity, resulting in down-regulation of gene expression. With bovine aortic endothelial cells (BAECs) and a cell-free system, 1,2-NQ was found to covalently bind to CREB, and inhibit its DNA binding activity under conditions that were blocked by dithiothreitol. CRE-dependent luciferase activity and the down-regulation of Bcl-2 expression were suppressed by exposure of BAECs to 1,2-NQ. This phenomenon was not seen with the hydrocarbon, naphthalene, which lacks any electrophilic properties. The results indicate that CREB is a molecular target for 1,2-NQ which through irreversible binding, inhibits the function of this transcription factor.