Merck
Novel, orally effective cyanide antidotes.
Journal of medicinal chemistry (2007-11-28)
Herbert T Nagasawa, David J W Goon, Daune L Crankshaw, Robert Vince, Steven E Patterson
ABSTRACT
A series of prodrugs of 3-mercaptopyruvate (3-MP), the substrate for the enzyme 3-mercaptopyruvate/cyanide sulfurtransferase (3-MPST) that converts cyanide to the nontoxic thiocyanate, which are highly effective cyanide antidotes, have been developed. These prodrugs of 3-MP are unique in being not only orally bioavailable, but may be administered up to an hour prior to cyanide as a prophylactic agent and are both rapid- or slow-acting when given parenterally.