Merck
  • Transformation of D-(-)-ribose into a natural product-like scaffold via a Lewis acid catalyzed intramolecular hetero-Diels-Alder reaction.
Transformation of D-(-)-ribose into a natural product-like scaffold via a Lewis acid catalyzed intramolecular hetero-Diels-Alder reaction.
Nucleosides, nucleotides & nucleic acids (2007-12-11)
Roland Messer, Cyril A Fuhrer, Robert Häner
ABSTRACT
Starting from D-(-)-ribose, a tricyclic natural product-like scaffold suitable for combinatorial derivatization was synthesized via an intramolecular hetero-Diels-Alder reaction. Lithium perchlorate was found to enhance the reaction rate and, at the same time, had a pronounced influence on the chemoselectivity of the reaction. The stereochemical course of the reaction, however, was not influenced by the Lewis acid.
Sigma-Aldrich
Lithium perchlorate, 99.99% trace metals basis
Sigma-Aldrich
Lithium perchlorate, battery grade, dry, 99.99% trace metals basis
Sigma-Aldrich
Lithium perchlorate, purum p.a., ≥98.0% (calc. based on dry substance, T), powder
Sigma-Aldrich
Magnesium perchlorate, ACS reagent
Sigma-Aldrich
Lithium perchlorate, ACS reagent, ≥95.0%
Sigma-Aldrich
Titanium(IV) isopropoxide, packaged for use in deposition systems
Sigma-Aldrich
Titanium(IV) isopropoxide, ≥97.0%
Sigma-Aldrich
Titanium(IV) isopropoxide, 97%
Sigma-Aldrich
Titanium(IV) isopropoxide, 99.999% trace metals basis