Merck
  • Trimethylsilyl trifluoromethanesulfonate (TMSOTf) assisted facile deprotection of N,O-acetonides.
Trimethylsilyl trifluoromethanesulfonate (TMSOTf) assisted facile deprotection of N,O-acetonides.
The Journal of organic chemistry (2007-12-14)
Kevin W C Poon, Kimberly M Lovell, Kendra N Dresner, Apurba Datta
ABSTRACT
Employing TMSOTf as an easily available reagent, we have developed a mild and efficient method for the deprotection of both terminal and internal N,0-acetonide functionalities. Various regularly used protecting groups and common organic functional moieties were found to be unaffected by the described reaction conditions. In a few representative examples, the present method was also extended to deprotect acetonides obtained from 1,2-, and 1,3-terminal diols. The acetonide deprotection protocol described herein is expected to be a useful addition to the presently available methods for performing the above transformation.
Sigma-Aldrich
Trimethylsilyl trifluoromethanesulfonate, 99%
Sigma-Aldrich
Trimethylsilyl trifluoromethanesulfonate, purum, ≥98.0% (T)