Merck
  • Acanthosulfate, a sulfated hydroxyhydroquinone sesterterpenoid from the sponge Acanthodendrilla sp.
Acanthosulfate, a sulfated hydroxyhydroquinone sesterterpenoid from the sponge Acanthodendrilla sp.
Journal of natural products (2008-02-09)
Lyndon M West, D John Faulkner
ABSTRACT
The marine sponge Acanthodendrilla sp. contains the proteasome inhibitor acanthosulfate ( 4), a disulfated merosesterterpene having a scalarane-type skeleton. The structure of acanthosulfate ( 4), which possesses an unusual configuration, was elucidated by interpretation of spectroscopic data.
Sigma-Aldrich
1,2,4-Benzenetriol, ReagentPlus®, 99%