Merck
  • Antioxidant activity of the new thiosulfinate derivative, S-benzyl phenylmethanethiosulfinate, from Petiveria alliacea L.
Antioxidant activity of the new thiosulfinate derivative, S-benzyl phenylmethanethiosulfinate, from Petiveria alliacea L.
Organic & biomolecular chemistry (2008-03-11)
Youji Okada, Kaoru Tanaka, Eisuke Sato, Haruo Okajima
ABSTRACT
The antioxidant effects of the new thiosulfinate derivative, S-benzyl phenylmethanethiosulfinate (BPT), against the oxidation of cumene and methyl linoleate (ML) in chlorobenzene were studied in detail using HPLC. The results showed that BPT provided effective inhibition with a well-defined induction period under these oxidation conditions, and it was found that the stoichiometric factor (n), the number of peroxyl radicals trapped by one antioxidant molecule, of BPT is about 2. We then undertook a thorough investigation aimed at elucidating the active structural site of BPT. Various model compounds, such as diphenyl disulfide, dibenzyl disulfide, S-phenyl benzenethiosulfinate and S-ethyl phenylmethanethiosulfinate, were used which provided evidence that the benzylic hydrogen of BPT is mainly associated with the peroxyl radical scavenging. Moreover, we measured the rate constant for the reaction of BPT with peroxyl radicals derived from cumene and ML in chlorobenzene, and based on these measurements, BPT reacts with these peroxyl radicals with a rate constant of k(inh) = 8.6 x 10(3) and 6.2 x 10(4) M(-1) s(-1), respectively.
Product Description
Supelco
Cumene, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Cumene, 98%
Supelco
Cumene, analytical standard