Merck
  • Synthesis of N-substituted indole-3-carboxylic acid derivatives via Cu(I)-catalyzed intramolecular amination of aryl bromides.
Synthesis of N-substituted indole-3-carboxylic acid derivatives via Cu(I)-catalyzed intramolecular amination of aryl bromides.
The Journal of organic chemistry (2008-05-13)
Ferdinand S Melkonyan, Alexander V Karchava, Marina A Yurovskaya
ABSTRACT
A variety of N-alkylated and N-arylated derivatives of methyl 1 H-indole-3-carboxylate were synthesized efficiently via Ullmann-type intramolecular arylamination, using the CuI-K 3PO 4-DMF system. This catalytic amination procedure can be performed with good to high yields under mild conditions under an air atmosphere.