Merck
  • Pyrrole-2-carboxylic acid as a ligand for the Cu-catalyzed reactions of primary anilines with aryl halides.
Pyrrole-2-carboxylic acid as a ligand for the Cu-catalyzed reactions of primary anilines with aryl halides.
The Journal of organic chemistry (2008-06-12)
Ryan A Altman, Kevin W Anderson, Stephen L Buchwald
ABSTRACT
Pyrrole 2-carboxylic acid (L5) was found to be an effective ligand for the Cu-catalyzed monoarylation of anilines with aryl iodides and bromides. Under the reported conditions (10% CuI/20% L5/DMSO/K3PO 4/80-100 degrees C/20-24 h), a variety of useful functional groups were tolerated, and moderate to good yields of the diaryl amine products were obtained.
Sigma-Aldrich
Pyrrole-2-carboxylic acid, 99%